missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-iodopyridine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 104830-06-0
Molecular Formula C5H5IN2
Molecular Weight (g/mol) 220.01
InChI Key UUDNBWSHTUFGDQ-UHFFFAOYSA-N
Synonym 2-amino-3-iodopyridine, 3-iodopyrid-2-ylamine, 3-iodo-pyridin-2-ylamine, 3-iodo-2-pyridylamine, 2-pyridinamine, 3-iodo, iodo-aminopyridine, pubchem1265, 2-amino-3-iodpyridine, acmc-20a0df
PubChem CID 2763151
IUPAC Name 3-iodopyridin-2-amine
SMILES C1=CC(=C(N=C1)N)I
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC441480050
SDS View Documents
Thermo Scientific Chemicals
441480050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

104830-06-0
220.01
2-amino-3-iodopyridine, 3-iodopyrid-2-ylamine, 3-iodo-pyridin-2-ylamine, 3-iodo-2-pyridylamine, 2-pyridinamine, 3-iodo, iodo-aminopyridine, pubchem1265, 2-amino-3-iodpyridine, acmc-20a0df
3-iodopyridin-2-amine
C5H5IN2
UUDNBWSHTUFGDQ-UHFFFAOYSA-N
2763151
C1=CC(=C(N=C1)N)I

Specifications

104830-06-0
100.0
96% min. (GC)
C5H5IN2
2-amino-3-iodopyridine, 3-iodopyrid-2-ylamine, 3-iodo-pyridin-2-ylamine, 3-iodo-2-pyridylamine, 2-pyridinamine, 3-iodo, iodo-aminopyridine, pubchem1265, 2-amino-3-iodpyridine, acmc-20a0df
C1=CC(=C(N=C1)N)I
220.01
220.01
2-Amino-3-iodopyridine
96.0
Authentic
Glass bottle
5g
UUDNBWSHTUFGDQ-UHFFFAOYSA-N
3-iodopyridin-2-amine
2763151
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only