missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-hydroxypyridine, 98%

CAS: 16867-03-1 | C5H6N2O | 110.116 g/mol

Supplier:  Thermo Scientific Chemicals A1066518

 View more versions of this product

Catalog No. AAA1066518


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Amino-3-hydroxypyridine
166°C to 174°C
C5H6N2O
50 g
109868
BMTSZVZQNMNPCT-UHFFFAOYSA-N
2-aminopyridin-3-ol
28114
98%
16867-03-1
Odorless
MFCD00006317
UN2811
2-amino-3-hydroxypyridine, 2-amino-3-pyridinol, 3-pyridinol, 2-amino, 2-amino-3-pyridol, 3-hydroxy-2-pyridinamine, 2-amino-pyridin-3-ol, 2-amino-3-hydroxy pyridine, unii-p8740pew4b, 3-hydroxy-2-aminopyridine, pubchem2574
C1=CC(=C(N=C1)N)O
110.116
110.12

Chemical Identifiers

16867-03-1
110.116
BMTSZVZQNMNPCT-UHFFFAOYSA-N
28114
C1=CC(=C(N=C1)N)O
C5H6N2O
MFCD00006317
2-amino-3-hydroxypyridine, 2-amino-3-pyridinol, 3-pyridinol, 2-amino, 2-amino-3-pyridol, 3-hydroxy-2-pyridinamine, 2-amino-pyridin-3-ol, 2-amino-3-hydroxy pyridine, unii-p8740pew4b, 3-hydroxy-2-aminopyridine, pubchem2574
2-aminopyridin-3-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c

H301-H311-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 240-886-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only