missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-hydroxypyridine, 98%

Catalog No. AAA1066530
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1066530 250 g
AAA1066509 10 g
AAA1066518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1066530 Supplier Thermo Scientific Chemicals Supplier No. A1066530
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16867-03-1 | C5H6N2O | 110.116 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16867-03-1
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
MDL Number MFCD00006317
InChI Key BMTSZVZQNMNPCT-UHFFFAOYSA-N
Synonym 2-amino-3-hydroxypyridine, 2-amino-3-pyridinol, 3-pyridinol, 2-amino, 2-amino-3-pyridol, 3-hydroxy-2-pyridinamine, 2-amino-pyridin-3-ol, 2-amino-3-hydroxy pyridine, unii-p8740pew4b, 3-hydroxy-2-aminopyridine, pubchem2574
PubChem CID 28114
IUPAC Name 2-aminopyridin-3-ol
SMILES C1=CC(=C(N=C1)N)O

Specifications

Melting Point 166°C to 174°C
Odor Odorless
Quantity 250 g
UN Number UN2811
Beilstein 109868
Formula Weight 110.12
Percent Purity 98%
Chemical Name or Material 2-Amino-3-hydroxypyridine
Hazard Category H301-H311-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 240-886-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only