missing translation for 'onlineSavingsMsg'
Learn More

2'-Amino-3'-hydroxyacetophenone 98.0+%, TCI America™

Catalog No. p-7117370
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13211G 1 g
A13215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13211G Supplier TCI America Supplier No. A13211G
Only null left

Chemical Identifiers

CAS 4502-10-7
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00191301
InChI Key DIIASMSSGMRMQF-UHFFFAOYSA-N
PubChem CID 20591
IUPAC Name 1-(2-amino-3-hydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C(=CC=C1)O)N

Specifications

Melting Point 182°C
Color Yellow
Quantity 1 g
Formula Weight 151.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Amino-3′-hydroxyacetophenone
RTECSNumber AM5950000
TSCA No