missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Amino-3'-hydroxyacetophenone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 182°C |
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 151.17 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2′-Amino-3′-hydroxyacetophenone |
Safety and Handling
| RTECSNumber | AM5950000 |
| TSCA | No |