missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-cyanothiophene 98.0+%, TCI America™

Catalog No. A24885G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A24885G Supplier TCI America Supplier No. A24885G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4651-82-5
Molecular Formula C5H4N2S
Molecular Weight (g/mol) 124.16
MDL Number MFCD00706298
InChI Key XVGHZFWFGXDIOU-UHFFFAOYSA-N
Synonym 2-amino-3-cyanothiophene, 2-amino-3-thiophenecarbonitrile, 2-amino-thiophene-3-carbonitrile, 3-thiophenecarbonitrile,2-amino, 3-thiophenecarbonitrile, 2-amino, 2-aminothiophen-3-carbonitril, pubchem5222, aminothiophenecarbonitrile, 2-amino-3-thienyl cyanide, 2-azanylthiophene-3-carbonitrile
PubChem CID 736607
IUPAC Name 2-aminothiophene-3-carbonitrile
SMILES NC1=C(C=CS1)C#N

Specifications

Melting Point 106°C
Color Yellow
Quantity 5 g
Formula Weight 124.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-cyanothiophene