missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-chloropyridine, 95%

Catalog No. AAH3016606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3016606 5 g
AAH3016603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3016606 Supplier Thermo Scientific Chemicals Supplier No. H3016606
Only null left

CAS: 39620-04-7 | C5H5ClN2 | 128.559 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39620-04-7
Molecular Formula C5H5ClN2
Molecular Weight (g/mol) 128.559
MDL Number MFCD03541052
InChI Key RZJPBQGRCNJYBU-UHFFFAOYSA-N
Synonym 2-amino-3-chloropyridine, 3-chloro-pyridin-2-ylamine, 2-amino-3-chloro pyridine, 3-chloro-2-pyridinamine, 3-chloro-2-pyridinylamine, 2-pyridinamine, 3-chloro, 3-chloro-2-pyridylamine, 2-amino-3-chloro-pyridine, 3-chloranylpyridin-2-amine, o-aminochloropyridine
PubChem CID 693267
IUPAC Name 3-chloropyridin-2-amine
SMILES C1=CC(=C(N=C1)N)Cl

Specifications

Melting Point 59°C to 62°C
Quantity 5 g
Formula Weight 128.56
Percent Purity 95%
Chemical Name or Material 2-Amino-3-chloropyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only