missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-chloro-5-methylpyridine 98.0+%, TCI America™

Catalog No. A18871G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18871G 1 g
A18875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A18871G Supplier TCI America Supplier No. A18871G
Only null left

Chemical Identifiers

CAS 31430-41-8
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.586
InChI Key COTHZSSUQJRBQH-UHFFFAOYSA-N
Synonym 6-Amino-5-chloro-3-picoline
PubChem CID 12251011
IUPAC Name 3-chloro-5-methylpyridin-2-amine
SMILES CC1=CC(=C(N=C1)N)Cl

Specifications

Melting Point 60°C
Quantity 1 g
Formula Weight 142.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-chloro-5-methylpyridine