missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-bromo-6-methylpyridine, 95%, Thermo Scientific Chemicals

$353.05 - $353.05

Chemical Identifiers

CAS 126325-46-0
Molecular Formula C6H8BrN2
Molecular Weight (g/mol) 188.05
MDL Number MFCD03095215
InChI Key JYWWGZAAXTYNRN-UHFFFAOYSA-O
Synonym 2-amino-3-bromo-6-methylpyridine, 2-amino-3-bromo-6-picoline, 6-amino-5-bromo-2-picoline, 2-pyridinamine, 3-bromo-6-methyl, 3-bromo-6-methyl-2-pyridylamine, 2-amine-3-bromo-6-methylpyridine, 3-bromo-6-methyl-pyridin-2-amine, 2-amino-3-bromo-6-picoline 2-amino-3-bromo-6-methylpyridine, pubchem1235, 6-amino-5-bromo-picoline
PubChem CID 2734416
IUPAC Name 2-amino-3-bromo-6-methylpyridin-1-ium
SMILES CC1=CC=C(Br)C(N)=[NH+]1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6616206
View Documents
Thermo Scientific Chemicals
H6616206
5 g
Each for $353.05
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

126325-46-0
188.05
JYWWGZAAXTYNRN-UHFFFAOYSA-O
2734416
CC1=CC=C(Br)C(N)=[NH+]1
C6H8BrN2
MFCD03095215
2-amino-3-bromo-6-methylpyridine, 2-amino-3-bromo-6-picoline, 6-amino-5-bromo-2-picoline, 2-pyridinamine, 3-bromo-6-methyl, 3-bromo-6-methyl-2-pyridylamine, 2-amine-3-bromo-6-methylpyridine, 3-bromo-6-methyl-pyridin-2-amine, 2-amino-3-bromo-6-picoline 2-amino-3-bromo-6-methylpyridine, pubchem1235, 6-amino-5-bromo-picoline
2-amino-3-bromo-6-methylpyridin-1-ium

Specifications

126325-46-0
White
MFCD03095215
UN2811
Light sensitive
JYWWGZAAXTYNRN-UHFFFAOYSA-O
2-amino-3-bromo-6-methylpyridin-1-ium
2734416
95%
2-Amino-3-bromo-6-methylpyridine
78°C to 83°C
C6H8BrN2
5 g
6190026
2-amino-3-bromo-6-methylpyridine, 2-amino-3-bromo-6-picoline, 6-amino-5-bromo-2-picoline, 2-pyridinamine, 3-bromo-6-methyl, 3-bromo-6-methyl-2-pyridylamine, 2-amine-3-bromo-6-methylpyridine, 3-bromo-6-methyl-pyridin-2-amine, 2-amino-3-bromo-6-picoline 2-amino-3-bromo-6-methylpyridine, pubchem1235, 6-amino-5-bromo-picoline
CC1=CC=C(Br)C(N)=[NH+]1
188.05
187.04
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only