missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-3-benzyloxypyridine 98.0+%, TCI America™
$65.24 - $193.86
Chemical Identifiers
| CAS | 24016-03-3 |
|---|---|
| Molecular Formula | C12H12N2O |
| Molecular Weight (g/mol) | 200.24 |
| MDL Number | MFCD00006316 |
| InChI Key | NMCBWICNRJLKKM-UHFFFAOYSA-N |
| Synonym | 2-amino-3-benzyloxypyridine, 3-benzyloxy pyridin-2-amine, 3-benzyloxy-2-pyridylamine, 2-pyridinamine, 3-phenylmethoxy, 2-amino-3-benzyloxy pyridine, 3ip, 3-phenylmethoxy-2-pyridylamine, 3-benzyloxy pyridin-2-amin, 3fty |
| PubChem CID | 90334 |
| IUPAC Name | 3-(benzyloxy)pyridin-2-amine |
| SMILES | NC1=NC=CC=C1OCC1=CC=CC=C1 |
Chemical Identifiers
| 24016-03-3 | |
| 200.24 | |
| NMCBWICNRJLKKM-UHFFFAOYSA-N | |
| 90334 | |
| NC1=NC=CC=C1OCC1=CC=CC=C1 |
| C12H12N2O | |
| MFCD00006316 | |
| 2-amino-3-benzyloxypyridine, 3-benzyloxy pyridin-2-amine, 3-benzyloxy-2-pyridylamine, 2-pyridinamine, 3-phenylmethoxy, 2-amino-3-benzyloxy pyridine, 3ip, 3-phenylmethoxy-2-pyridylamine, 3-benzyloxy pyridin-2-amin, 3fty | |
| 3-(benzyloxy)pyridin-2-amine |
Specifications
| 24016-03-3 | |
| Yellow | |
| MFCD00006316 | |
| 2-amino-3-benzyloxypyridine, 3-benzyloxy pyridin-2-amine, 3-benzyloxy-2-pyridylamine, 2-pyridinamine, 3-phenylmethoxy, 2-amino-3-benzyloxy pyridine, 3ip, 3-phenylmethoxy-2-pyridylamine, 3-benzyloxy pyridin-2-amin, 3fty | |
| NC1=NC=CC=C1OCC1=CC=CC=C1 | |
| 200.24 | |
| 200.24 | |
| Crystalline Powder |
| 97°C | |
| C12H12N2O | |
| 5 g | |
| NMCBWICNRJLKKM-UHFFFAOYSA-N | |
| 3-(benzyloxy)pyridin-2-amine | |
| 90334 | |
| ≥98.0% (T) | |
| 2-Amino-3-benzyloxypyridine |
Safety and Handling
TSCA : No