missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-benzyloxypyridine 98.0+%, TCI America™

Catalog No. A20805G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A20805G 5 g
A208025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A20805G Supplier TCI America Supplier No. A20805G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 24016-03-3 | C12H12N2O | 200.24 g/mol

Chemical Identifiers

CAS 24016-03-3
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.24
MDL Number MFCD00006316
InChI Key NMCBWICNRJLKKM-UHFFFAOYSA-N
Synonym 2-amino-3-benzyloxypyridine, 3-benzyloxy pyridin-2-amine, 3-benzyloxy-2-pyridylamine, 2-pyridinamine, 3-phenylmethoxy, 2-amino-3-benzyloxy pyridine, 3ip, 3-phenylmethoxy-2-pyridylamine, 3-benzyloxy pyridin-2-amin, 3fty
PubChem CID 90334
IUPAC Name 3-(benzyloxy)pyridin-2-amine
SMILES NC1=NC=CC=C1OCC1=CC=CC=C1

Specifications

Melting Point 97°C
Color Yellow
Quantity 5 g
Formula Weight 200.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-benzyloxypyridine