missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3-benzyloxypyrazine, 96%

Catalog No. AAH63162MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH63162MD 250 mg
AAH6316203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH63162MD Supplier Thermo Scientific Chemicals Supplier No. H63162MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 110223-15-9 | C11H11N3O | 201.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110223-15-9
Molecular Formula C11H11N3O
Molecular Weight (g/mol) 201.23
MDL Number MFCD09838954
InChI Key QKEQFJXWHGVJJU-UHFFFAOYSA-N
Synonym 2-amino-3-benzyloxypyrazine, 3-benzyloxy pyrazin-2-amine, 2-pyrazinamine,3-phenylmethoxy, 2-pyrazinamine, 3-phenylmethoxy, zlchem 177, pubchem19570, acmc-20a0ri, 2-aminol-3-benzyloxypyrazine, 3-phenylmethoxy-2-pyrazinamine
PubChem CID 13900234
IUPAC Name 3-phenylmethoxypyrazin-2-amine
SMILES NC1=NC=CN=C1OCC1=CC=CC=C1

Specifications

Quantity 250 mg
Formula Weight 201.22
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Amino-3-benzyloxypyrazine
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only