missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dichlorobenzonitrile 98.0+%, TCI America™

Catalog No. A30621G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30621G 1 g
A30625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30621G Supplier TCI America Supplier No. A30621G
Only null left

Chemical Identifiers

CAS 36764-94-0
Molecular Formula C7H4Cl2N2
Molecular Weight (g/mol) 187.02
MDL Number MFCD00158947
InChI Key ZHKNDJRPOVUPMT-UHFFFAOYSA-N
Synonym 2-amino-3,5-dichloro-benzonitrile, 2-amino-3,5-dichlorobenzenecarbonitrile, pubchem11850, 3,5-dichloroanthranilonitrile, buttpark 52\14-89, benzonitrile,2-amino-3,5-dichloro, 2-amino-3,5-dichlorobenzonitrile
PubChem CID 737215
IUPAC Name 2-amino-3,5-dichlorobenzonitrile
SMILES NC1=C(Cl)C=C(Cl)C=C1C#N

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 1 g
Formula Weight 187.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3,5-dichlorobenzonitrile