missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dichloro-6-methylpyridine 97.0+%, TCI America™

Catalog No. p-7117640
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
A19331G 1 g
A19335G 5 g
2 options

Catalog No. A19331G

Supplier: TCI America A19331G

Only null left

Chemical Identifiers

CAS 22137-52-6
Molecular Formula C6H6Cl2N2
Molecular Weight (g/mol) 177.028
MDL Number MFCD00129029
InChI Key NQYNSMVHXHIGHZ-UHFFFAOYSA-N
Synonym 6-Amino-3,5-dichloro-2-picoline
PubChem CID 1415863
IUPAC Name 3,5-dichloro-6-methylpyridin-2-amine
SMILES CC1=NC(=C(C=C1Cl)Cl)N

Specifications

Melting Point 134°C
Quantity 1 g
Formula Weight 177.03
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3,5-dichloro-6-methylpyridine