missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-3,5-dibromopyrazine 98.0+%, TCI America™

Catalog No. A18061G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18061G 1 g
A18065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A18061G Supplier TCI America Supplier No. A18061G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24241-18-7
Molecular Formula C4H3Br2N3
Molecular Weight (g/mol) 252.90
MDL Number MFCD00673150
InChI Key DTLBKXRFWUERQN-UHFFFAOYSA-N
Synonym 2-amino-3,5-dibromopyrazine, 3,5-dibromo-2-pyrazinamine, 3,5-dibromopyrazin-2-ylamine, 3,5-dibromopyrazine-2-ylamine, amino-3,5-dibromopyrazine, pyrazinamine, 3,5-dibromo, 3,5-dibromopyrazine-2-amine, 2-amino-3,5-dibromo pyrazine, 2-amino-3,5-dibromo-pyrazine, 2-pyrazinamine, 3,5-dibromo
PubChem CID 620004
IUPAC Name 3,5-dibromopyrazin-2-amine
SMILES NC1=NC=C(Br)N=C1Br

Specifications

Melting Point 114°C
Quantity 1 g
Formula Weight 252.90
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3,5-dibromopyrazine