missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-methyl-1,3-propanediol 98.0+%, TCI America™

Catalog No. p-7116841
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
A033225G 25 g
A0332100G 100 g
A0332500G 500 g
3 options

Catalog No. A033225G

Supplier: TCI America A033225G

Only null left

Chemical Identifiers

CAS 115-69-5
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.137
MDL Number MFCD00004678
InChI Key UXFQFBNBSPQBJW-UHFFFAOYSA-N
Synonym 2-amino-2-methyl-1,3-propanediol, ampd, aminoglycol, gentimon, isobutandiol-2-amine, aminomethyl propanediol, ammediol, 1,1-di hydroxymethyl ethylamine, 1,3-propanediol, 2-amino-2-methyl, 2-amino-2-methyl-1,3-propandiol
PubChem CID 1531
ChEBI CHEBI:991
IUPAC Name 2-amino-2-methylpropane-1,3-diol
SMILES CC(CO)(CO)N

Specifications

Melting Point 108°C
Color White-Yellow
Boiling Point 152°C
Quantity 25 g
Formula Weight 105.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-2-methyl-1,3-propanediol
EINECSNumber (2)-0321
RTECSNumber TY2975000
TSCA Yes