missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-2-methyl-1,3-propanediol, 99+%

Catalog No. aaj6314422
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
250 g
Catalog No. Quantity
AAJ6314422 100 g
AAJ6314414 25 g
AAJ6314430 250 g
3 options

Catalog No. AAJ6314422

Supplier: Thermo Scientific Chemicals J6314422

Only null left

A useful biological buffer | CAS: 115-69-5 | C4H11NO2 | 105.137 g/mol

A useful biological buffer2-Amino-2-methyl-1,3-propanediol is used to make surfactants, vulcanization accelerators and as a biological buffer. It acts as an absorbent for acidic gases and emulsifying agent for cosmetics, mineral oil, leather dressings, textile specialties and cleaning compounds. It is also used in hair sprays. It acts as a buffer for the determination of alkaline phosphatase activity and spacer in isotachophoresis of proteins. It serves as a reaction buffer for enzymes with an activity in the basic pH range such as alkaline phosphatase. Further, it is used in the preparation of 4-hydroxymethyl-4-methyl-oxazolidin-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A useful biological buffer2-Amino-2-methyl-1,3-propanediol is used to make surfactants, vulcanization accelerators and as a biological buffer. It acts as an absorbent for acidic gases and emulsifying agent for cosmetics, mineral oil, leather dressings, textile specialties and cleaning compounds. It is also used in hair sprays. It acts as a buffer for the determination of alkaline phosphatase activity and spacer in isotachophoresis of proteins. It serves as a reaction buffer for enzymes with an activity in the basic pH range such as alkaline phosphatase. Further, it is used in the preparation of 4-hydroxymethyl-4-methyl-oxazolidin-2-one.

Solubility
Soluble in water and other organic solvents.

Notes
Hygroscopic. Air and moisture sensitive. Incompatible with strong oxidizing agents, aluminum, copper, brass and strong acids.

Chemical Identifiers

CAS 115-69-5
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.137
MDL Number MFCD00004678
InChI Key UXFQFBNBSPQBJW-UHFFFAOYSA-N
Synonym 2-amino-2-methyl-1,3-propanediol, ampd, aminoglycol, gentimon, isobutandiol-2-amine, aminomethyl propanediol, ammediol, 1,1-di hydroxymethyl ethylamine, 1,3-propanediol, 2-amino-2-methyl, 2-amino-2-methyl-1,3-propandiol
PubChem CID 1531
ChEBI CHEBI:991
IUPAC Name 2-amino-2-methylpropane-1,3-diol
SMILES CC(CO)(CO)N

Specifications

Melting Point 100°C to 110°C
Color White
Boiling Point 151°C (10 mmHg)
Linear Formula (HOCH2)2C(NH2)CH3
Quantity 100 g
Beilstein 635708
Merck Index 14,448
Solubility Information Soluble in water and other organic solvents.
Formula Weight 105.14
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material 2-Amino-2-methyl-1,3-propanediol
Show More Show Less
Hazard Category H290-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P234-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 204-100-7
RTECSNumber TY2975000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only