missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Amino-1-propanol, 98%

Catalog No. aal0360806
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAL0360806 5 g
AAL0360814 25 g
2 options

Catalog No. AAL0360806

Supplier: Thermo Scientific Chemicals L0360806

Only null left

CAS: 6168-72-5 | C3H9NO | 75.11 g/mol

DL-Alaninol has been used in the synthesis of N6-α-(I-hydroxypropyl) lysine, diastereomers of DL-β-amino alcohols, enantiopure and racemic samples of 2-methyl-N-tosylaziridine, (±)-3-(5-dimethylcarbamoyl-pent-1-enyl)-N-(2-hydroxy-1-methyl-ethyl)benzamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Alaninol has been used in the synthesis of N6-α-(I-hydroxypropyl) lysine, diastereomers of DL-β-amino alcohols, enantiopure and racemic samples of 2-methyl-N-tosylaziridine, (±)-3-(5-dimethylcarbamoyl-pent-1-enyl)-N-(2-hydroxy-1-methyl-ethyl)benzamide.

Solubility
Fully miscible in water.

Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, air, water/ moisture.

Chemical Identifiers

CAS 6168-72-5
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00008085
InChI Key BKMMTJMQCTUHRP-UHFFFAOYNA-N
Synonym dl-alaninol, 2-aminopropanol, 2-amino-1-propanol, dl-2-amino-1-propanol, 1-propanol, 2-amino, 1-hydroxy-2-aminopropane, 2-amino-2-methylethanol, alaninol, propanol, 2-amino, 1-methyl-2-hydroxyethylamine
PubChem CID 5126
IUPAC Name 2-aminopropan-1-ol
SMILES CC(N)CO

Specifications

Melting Point 8°C
Density 0.965
Boiling Point 174°C to 176°C
Flash Point 83°C (181°F)
Refractive Index 1.4495
Quantity 5 g
UN Number UN2735
Beilstein 1209234
Sensitivity Air sensitive; Hygroscopic
Solubility Information Fully miscible in water.
Formula Weight 75.11
Percent Purity 98%
Chemical Name or Material (+/-)-2-Amino-1-propanol
Show More Show Less
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 228-207-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only