missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-1-phenylethanol 97.0+%, TCI America™
$389.59 - $389.59
Chemical Identifiers
| CAS | 7568-93-6 |
|---|---|
| Molecular Formula | C8H11NO |
| Molecular Weight (g/mol) | 137.18 |
| MDL Number | MFCD00008137 |
| InChI Key | ULSIYEODSMZIPX-UHFFFAOYNA-N |
| Synonym | phenylethanolamine, 2-hydroxy-2-phenylethylamine, phenethanolamine, beta-hydroxyphenethylamine, 2-phenyl-2-hydroxyethylamine, bisnorephedrine, beta-phenethanolamine, alpha-aminomethyl benzyl alcohol, 2-amino-1-phenylethan-1-ol, 2-amino-1-phenyl-1-ethanol |
| PubChem CID | 1000 |
| ChEBI | CHEBI:16343 |
| IUPAC Name | 2-amino-1-phenylethan-1-ol |
| SMILES | NCC(O)C1=CC=CC=C1 |
Chemical Identifiers
| 7568-93-6 | |
| 137.18 | |
| ULSIYEODSMZIPX-UHFFFAOYNA-N | |
| 1000 | |
| 2-amino-1-phenylethan-1-ol |
| C8H11NO | |
| MFCD00008137 | |
| phenylethanolamine, 2-hydroxy-2-phenylethylamine, phenethanolamine, beta-hydroxyphenethylamine, 2-phenyl-2-hydroxyethylamine, bisnorephedrine, beta-phenethanolamine, alpha-aminomethyl benzyl alcohol, 2-amino-1-phenylethan-1-ol, 2-amino-1-phenyl-1-ethanol | |
| CHEBI:16343 | |
| NCC(O)C1=CC=CC=C1 |
Specifications
| 7568-93-6 | |
| White-Yellow | |
| MFCD00008137 | |
| 3259 | |
| ULSIYEODSMZIPX-UHFFFAOYNA-N | |
| 2-amino-1-phenylethan-1-ol | |
| 1000 | |
| 137.18 | |
| Crystalline Powder |
| 57°C | |
| C8H11NO | |
| 5 g | |
| phenylethanolamine, 2-hydroxy-2-phenylethylamine, phenethanolamine, beta-hydroxyphenethylamine, 2-phenyl-2-hydroxyethylamine, bisnorephedrine, beta-phenethanolamine, alpha-aminomethyl benzyl alcohol, 2-amino-1-phenylethan-1-ol, 2-amino-1-phenyl-1-ethanol | |
| NCC(O)C1=CC=CC=C1 | |
| 137.18 | |
| CHEBI:16343 | |
| ≥97.0% (GC,T) | |
| 2-Amino-1-phenylethanol |
Safety and Handling
RTECSNumber : DN5500000
TSCA : Yes