missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1-phenylethanol 97.0+%, TCI America™

Catalog No. A10185G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A10185G Supplier TCI America Supplier No. A10185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7568-93-6
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00008137
InChI Key ULSIYEODSMZIPX-UHFFFAOYNA-N
Synonym phenylethanolamine, 2-hydroxy-2-phenylethylamine, phenethanolamine, beta-hydroxyphenethylamine, 2-phenyl-2-hydroxyethylamine, bisnorephedrine, beta-phenethanolamine, alpha-aminomethyl benzyl alcohol, 2-amino-1-phenylethan-1-ol, 2-amino-1-phenyl-1-ethanol
PubChem CID 1000
ChEBI CHEBI:16343
IUPAC Name 2-amino-1-phenylethan-1-ol
SMILES NCC(O)C1=CC=CC=C1

Specifications

Melting Point 57°C
Color White-Yellow
Quantity 5 g
UN Number 3259
Formula Weight 137.18
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-1-phenylethanol
RTECSNumber DN5500000
TSCA Yes