Learn More
2-Amino-1-methylbenzimidazole, 99+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 276300100
| Quantity | 10g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C8H9N3 | |
| MFCD00142855 | |
| 2-amino-1-methylbenzimidazole, 1-methyl-1h-benzimidazol-2-amine, 1-methyl-1h-benzo d imidazol-2-amine, 1-methyl-2-aminobenzimidazole, 1h-benzimidazol-2-amine, 1-methyl, 1-methyl-1h-1,3-benzodiazol-2-amine, ccris 4355, 2-amino-1-methyl-benzimidazole, 1h-benzimidazol-2-amine,1-methyl, 1-methyl-1h-benzoimidazol-2-ylamine | |
| 1-methylbenzimidazol-2-amine |
Specifications
| 2-Amino-1-methylbenzimidazole | |
| 99.0 | |
| Authentic | |
| Glass bottle | |
| 10g | |
| 2-amino-1-methylbenzimidazole, 1-methyl-1h-benzimidazol-2-amine, 1-methyl-1h-benzo d imidazol-2-amine, 1-methyl-2-aminobenzimidazole, 1h-benzimidazol-2-amine, 1-methyl, 1-methyl-1h-1,3-benzodiazol-2-amine, ccris 4355, 2-amino-1-methyl-benzimidazole, 1h-benzimidazol-2-amine,1-methyl, 1-methyl-1h-benzoimidazol-2-ylamine | |
| CN1C2=CC=CC=C2N=C1N | |
| 147.18 | |
| 147.18 |
| 1622-57-7 | |
| 100.0 | |
| 99% min. (GC) | |
| C8H9N3 | |
| MFCD00142855 | |
| XDFZKQJLNGNJAN-UHFFFAOYSA-N | |
| 1-methylbenzimidazol-2-amine | |
| 74187 | |
| 99+% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective glov
GHS Signal Word: Warning