missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1-methoxybutane 90.0+%, TCI America™

Catalog No. M0746500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0746500ML 500 mL
M074625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0746500ML Supplier TCI America Supplier No. M0746500ML
Only null left

Chemical Identifiers

CAS 63448-63-5
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.165
MDL Number MFCD00047920
InChI Key QAFODUGVXFNLBE-UHFFFAOYSA-N
Synonym 1-Methoxy-sec-butylamine
PubChem CID 93837
IUPAC Name 1-methoxybutan-2-amine
SMILES CCC(COC)N

Specifications

Color Yellow
Quantity 500 mL
UN Number 2734
Formula Weight 103.17
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Amino-1-methoxybutane