missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-1-methoxybutane 90.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 mL |
| UN Number | 2734 |
| Formula Weight | 103.17 |
| Percent Purity | ≥90.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Amino-1-methoxybutane |
Safety and Handling
| TSCA | Yes |