missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Amino-1-butanol, 97%

Catalog No. AAB24398AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB24398AP 500 mL
AAB24398AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB24398AP Supplier Thermo Scientific Chemicals Supplier No. B24398AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 96-20-8 | C4H11NO | 89.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 96-20-8
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.138
MDL Number MFCD00008095
InChI Key JCBPETKZIGVZRE-UHFFFAOYSA-N
Synonym 2-amino-1-butanol, 2-aminobutanol, dl-2-amino-1-butanol, 1-butanol, 2-amino, 2-aminobutyl alcohol, 1-hydroxy-2-butylamine, butanol-2-amine, 1-hydroxymethyl propylamine, 2-amino-1-hydroxybutane, 2-amino-n-butyl alcohol
PubChem CID 22129
IUPAC Name 2-aminobutan-1-ol
SMILES CCC(CO)N

Specifications

Melting Point -2°C
Density 0.944
pH 11.1
Boiling Point 176°C to 178°C
Flash Point 89°C (192°F)
Odor Amine-like
Refractive Index 1.4525
Quantity 500 mL
UN Number UN2735
Beilstein 1098274
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,427
Formula Weight 89.14
Percent Purity 97%
Chemical Name or Material (+/-)-2-Amino-1-butanol
Show More Show Less
Hazard Category H227-H290-H302-H312-H314-H332-H335
Hazard Statement GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P233-P234-P240-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P374-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 202-488-2
RTECSNumber EK9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only