missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1-(4-nitrophenyl)ethan-1-one hydrochloride hydrate, 97%, Thermo Scientific™

Catalog No. BTB13907CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BTB13907CB 250mg Amber Glass Bottle
BTB13907DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB13907CB Supplier Thermo Scientific Chemicals Supplier No. BTB13907CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5425-81-0
Molecular Formula C8H9ClN2O3
Molecular Weight (g/mol) 216.621
MDL Number MFCD00025591
InChI Key UJBOVJRECBNSDI-UHFFFAOYSA-N
Synonym 2-amino-1-4-nitrophenyl ethanone hydrochloride, 2-amino-4'-nitro acetophenone hydrochloride, 2-amino-p-nitro-acetophenone hcl, 2-amino-p-nitro-acetophenone hydrochloride, 2-amino-1-4-nitro-phenyl ethanone hydrochloride, 2-amino-1-4-nitro-phenyl-ethanone hydrochloride, ethanone, 2-amino-1-4-nitrophenyl-, monohydrochloride, 2-amino-1-4-nitrophenyl ethan-1-one, chloride, 4-nitrophenacylamine hydrochloride hydrate, aronis24405
PubChem CID 2792801
IUPAC Name 2-amino-1-(4-nitrophenyl)ethanone;hydrochloride
SMILES C1=CC(=CC=C1C(=O)CN)[N+](=O)[O-].Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula HCl·H2O
Quantity 250mg
Formula Weight 234.64
Percent Purity 97%
Chemical Name or Material 2-Amino-1-(4-nitrophenyl)ethan-1-one hydrochloride hydrate
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.