missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1,3,4-thiadiazole 98.0+%, TCI America™

$50.05 - $117.44

Chemical Identifiers

CAS 4005-51-0
Molecular Formula C2H3N3S
Molecular Weight (g/mol) 101.127
MDL Number MFCD00003107
InChI Key QUKGLNCXGVWCJX-UHFFFAOYSA-N
Synonym 2-amino-1,3,4-thiadiazole, aminothiadiazole, 2-aminothiadiazole, atda, aminothiadazole, unii-6l41aok74p, 1,3,4-thiadiazole, 2-amino, 1,3,4 thiadiazol-2-ylamine, 1,3,4-thiadiazol-2-ylamine
PubChem CID 19909
IUPAC Name 1,3,4-thiadiazol-2-amine
SMILES C1=NN=C(S1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A10605G
View Documents
TCI America
A10605G
5 g
Each for $50.05
Only null left
 
A106025G
View Documents
TCI America
A106025G
25 g
Each for $117.44
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4005-51-0
101.127
QUKGLNCXGVWCJX-UHFFFAOYSA-N
19909
C1=NN=C(S1)N
C2H3N3S
MFCD00003107
2-amino-1,3,4-thiadiazole, aminothiadiazole, 2-aminothiadiazole, atda, aminothiadazole, unii-6l41aok74p, 1,3,4-thiadiazole, 2-amino, 1,3,4 thiadiazol-2-ylamine, 1,3,4-thiadiazol-2-ylamine
1,3,4-thiadiazol-2-amine

Specifications

4005-51-0
White-Yellow
MFCD00003107
2811
QUKGLNCXGVWCJX-UHFFFAOYSA-N
1,3,4-thiadiazol-2-amine
19909
≥98.0% (T)
2-Amino-1,3,4-thiadiazole
183°C
C2H3N3S
5 g
2-amino-1,3,4-thiadiazole, aminothiadiazole, 2-aminothiadiazole, atda, aminothiadazole, unii-6l41aok74p, 1,3,4-thiadiazole, 2-amino, 1,3,4 thiadiazol-2-ylamine, 1,3,4-thiadiazol-2-ylamine
C1=NN=C(S1)N
101.127
101.13
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : XI3000000

TSCA : No

SDS
Documents

Documents