missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-1,3,4-thiadiazole, 98+%

Catalog No. p-7043546
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0124406 5 g
AAL0124414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0124406 Supplier Thermo Scientific Chemicals Supplier No. L0124406
Only null left

CAS: 4005-51-0 | C2H3N3S | 101.127 g/mol

2-Amino-1,3,4-thiadiazole, is used as an intermediate for ceftraoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-1,3,4-thiadiazole, is used as an intermediate for ceftraoline.

Solubility
It is soluble in water, hot methanol- very faint turbidity.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 4005-51-0
Molecular Formula C2H3N3S
Molecular Weight (g/mol) 101.127
MDL Number MFCD00003107
InChI Key QUKGLNCXGVWCJX-UHFFFAOYSA-N
Synonym 2-amino-1,3,4-thiadiazole, aminothiadiazole, 2-aminothiadiazole, atda, aminothiadazole, unii-6l41aok74p, 1,3,4-thiadiazole, 2-amino, 1,3,4 thiadiazol-2-ylamine, 1,3,4-thiadiazol-2-ylamine
PubChem CID 19909
IUPAC Name 1,3,4-thiadiazol-2-amine
SMILES C1=NN=C(S1)N

Specifications

Melting Point 188°C to 194°C
Quantity 5 g
UN Number UN2811
Beilstein 107135
Solubility Information It is soluble in water,hot methanol- very faint turbidity.
Formula Weight 101.13
Percent Purity ≥98%
Chemical Name or Material 2-Amino-1,3,4-thiadiazole
Hazard Category H301-H312+H332-H315-H319-H335-H341-H361d-H371
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 223-657-7
RTECSNumber XI3000000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only