missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-1,3,4-thiadiazole, 98+%
Description
2-Amino-1,3,4-thiadiazole, is used as an intermediate for ceftraoline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-1,3,4-thiadiazole, is used as an intermediate for ceftraoline.
Solubility
It is soluble in water, hot methanol- very faint turbidity.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
2-Amino-1,3,4-thiadiazole, is used as an intermediate for ceftraoline.
Solubility
It is soluble in water, hot methanol- very faint turbidity.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
Specifications
| Melting Point | 188°C to 194°C |
| Quantity | 25 g |
| UN Number | UN2811 |
| Beilstein | 107135 |
| Solubility Information | It is soluble in water,hot methanol- very faint turbidity. |
| Formula Weight | 101.13 |
| Percent Purity | ≥98% |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Amino-1,3,4-thiadiazole |
Safety and Handling
| Hazard Category | H301-H312+H332-H315-H319-H335-H341-H361d-H371 |
| Hazard Statement | GHS H StatementH301-H315-H319-H335 Toxic if swallowed.Causes skin irritation.Causes serious eye irritation.May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 223-657-7 |
| RTECSNumber | XI3000000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only