missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Allyloxybenzaldehyde 96.0+%, TCI America™
$154.02 - $610.28
Chemical Identifiers
| CAS | 28752-82-1 |
|---|---|
| Molecular Formula | C10H10O2 |
| Molecular Weight (g/mol) | 162.188 |
| MDL Number | MFCD00014130 |
| InChI Key | BXCJDECTRRMSCV-UHFFFAOYSA-N |
| Synonym | 2-allyloxy benzaldehyde, 2-allyloxybenzaldehyde, o-allyloxy benzaldehyde, o-allyloxybenzaldehyde, benzaldehyde, 2-2-propenyloxy, benzaldehyde, o-allyloxy, 2-prop-2-en-1-yloxy benzaldehyde, 2-prop-2-enyloxybenzaldehyde, allylsalicylaldehyde, o-allylsalicylaldehyde |
| PubChem CID | 101335 |
| IUPAC Name | 2-prop-2-enoxybenzaldehyde |
| SMILES | C=CCOC1=CC=CC=C1C=O |
Chemical Identifiers
| 28752-82-1 | |
| 162.188 | |
| BXCJDECTRRMSCV-UHFFFAOYSA-N | |
| 101335 | |
| C=CCOC1=CC=CC=C1C=O |
| C10H10O2 | |
| MFCD00014130 | |
| 2-allyloxy benzaldehyde, 2-allyloxybenzaldehyde, o-allyloxy benzaldehyde, o-allyloxybenzaldehyde, benzaldehyde, 2-2-propenyloxy, benzaldehyde, o-allyloxy, 2-prop-2-en-1-yloxy benzaldehyde, 2-prop-2-enyloxybenzaldehyde, allylsalicylaldehyde, o-allylsalicylaldehyde | |
| 2-prop-2-enoxybenzaldehyde |
Specifications
| 28752-82-1 | |
| C10H10O2 | |
| 5 g | |
| BXCJDECTRRMSCV-UHFFFAOYSA-N | |
| 2-prop-2-enoxybenzaldehyde | |
| 101335 | |
| ≥96.0% (GC) | |
| 2-Allyloxybenzaldehyde |
| Yellow | |
| MFCD00014130 | |
| 2-allyloxy benzaldehyde, 2-allyloxybenzaldehyde, o-allyloxy benzaldehyde, o-allyloxybenzaldehyde, benzaldehyde, 2-2-propenyloxy, benzaldehyde, o-allyloxy, 2-prop-2-en-1-yloxy benzaldehyde, 2-prop-2-enyloxybenzaldehyde, allylsalicylaldehyde, o-allylsalicylaldehyde | |
| C=CCOC1=CC=CC=C1C=O | |
| 162.188 | |
| 162.19 | |
| Liquid |
Safety and Handling
TSCA : No