missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Allyloxybenzaldehyde 96.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 162.19 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Allyloxybenzaldehyde |
Safety and Handling
| TSCA | No |