missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Allyl-5,5-dimethyl-1,3,2-dioxaborinane (stabilized with Phenothiazine) 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Quantity | 5 g |
| UN Number | 1993 |
| Formula Weight | 154.02 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Allyl-5,5-dimethyl-1,3,2-dioxaborinane (stabilized with Phenothiazine) |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |