missing translation for 'onlineSavingsMsg'
Learn More

2-Allyl-5,5-dimethyl-1,3,2-dioxaborinane (stabilized with Phenothiazine) 98.0+%, TCI America™

Catalog No. A25741G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25741G 1 g
A25745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25741G Supplier TCI America Supplier No. A25741G
Only null left

Chemical Identifiers

CAS 911482-75-2
Molecular Formula C8H15BO2
Molecular Weight (g/mol) 154.02
MDL Number MFCD27966263
InChI Key SBZNRMJWIVCYBF-UHFFFAOYSA-N
PubChem CID 11851265
IUPAC Name 5,5-dimethyl-2-(prop-2-en-1-yl)-1,3,2-dioxaborinane
SMILES CC1(C)COB(CC=C)OC1

Specifications

Color Colorless
Quantity 1 g
UN Number 1993
Formula Weight 154.02
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Allyl-5,5-dimethyl-1,3,2-dioxaborinane (stabilized with Phenothiazine)
TSCA No
Recommended Storage Refrigerator