missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Adamantanone 98.0+%, TCI America™
$78.29 - $471.60
Chemical Identifiers
| CAS | 700-58-3 |
|---|---|
| Molecular Formula | C10H14O |
| Molecular Weight (g/mol) | 150.221 |
| MDL Number | MFCD00074737 |
| InChI Key | IYKFYARMMIESOX-UHFFFAOYSA-N |
| Synonym | 2-adamantanone, adamantanone, 2-adamantone, 2-oxoadamantane, tricyclo 3.3.1.13,7 decanone, unii-ui7w503l08, tricyclo 3,3,1,13,7 decanone, tricyclo 3.3.1.1 sup 3,7 decanone, tricyclo 3.3.1.13,7 decan-2-one |
| PubChem CID | 64151 |
| ChEBI | CHEBI:40611 |
| IUPAC Name | adamantan-2-one |
| SMILES | C1C2CC3CC1CC(C2)C3=O |
Chemical Identifiers
| 700-58-3 | |
| 150.221 | |
| IYKFYARMMIESOX-UHFFFAOYSA-N | |
| 64151 | |
| adamantan-2-one |
| C10H14O | |
| MFCD00074737 | |
| 2-adamantanone, adamantanone, 2-adamantone, 2-oxoadamantane, tricyclo 3.3.1.13,7 decanone, unii-ui7w503l08, tricyclo 3,3,1,13,7 decanone, tricyclo 3.3.1.1 sup 3,7 decanone, tricyclo 3.3.1.13,7 decan-2-one | |
| CHEBI:40611 | |
| C1C2CC3CC1CC(C2)C3=O |
Specifications
| 700-58-3 | |
| C10H14O | |
| 5 g | |
| IYKFYARMMIESOX-UHFFFAOYSA-N | |
| adamantan-2-one | |
| 64151 | |
| 150.22 | |
| Crystalline Powder |
| White | |
| MFCD00074737 | |
| 2-adamantanone, adamantanone, 2-adamantone, 2-oxoadamantane, tricyclo 3.3.1.13,7 decanone, unii-ui7w503l08, tricyclo 3,3,1,13,7 decanone, tricyclo 3.3.1.1 sup 3,7 decanone, tricyclo 3.3.1.13,7 decan-2-one | |
| C1C2CC3CC1CC(C2)C3=O | |
| 150.221 | |
| CHEBI:40611 | |
| ≥98.0% (GC) | |
| 2-Adamantanone |
Safety and Handling
EINECSNumber : (4)-1894
RTECSNumber : AU5018000
TSCA : No