missing translation for 'onlineSavingsMsg'
Learn More

2-Adamantanone Oxime 98.0+%, TCI America™

$206.90 - $533.13

Chemical Identifiers

CAS 4500-12-3
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.24
MDL Number MFCD00078273
InChI Key RABVIFXMFZFITE-KHPPLWFESA-N
Synonym 2-adamantanone oxime, adamantan-2-one oxime, adamantanone oxime, 2-adamantanone, oxime, 2-hydroxyiminoadamantane, adamantanone-2-oxime, 2-isonitrosoadamantane, 2-hydroxyimino adamantane, n-2-adamantylidene hydroxylamine, n-adamantan-2-ylidene hydroxylamine
PubChem CID 64158
IUPAC Name N-(2-adamantylidene)hydroxylamine
SMILES C1C2CC3CC1CC(C2)C3=NO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A21105G
View Documents
TCI America
A21105G
5 g
Each for $206.90
Only null left
 
A211025G
View Documents
TCI America
A211025G
25 g
Each for $533.13
Only null left
 
Description

Description

Specifications

Chemical Identifiers

4500-12-3
165.24
RABVIFXMFZFITE-KHPPLWFESA-N
64158
C1C2CC3CC1CC(C2)C3=NO
C10H15NO
MFCD00078273
2-adamantanone oxime, adamantan-2-one oxime, adamantanone oxime, 2-adamantanone, oxime, 2-hydroxyiminoadamantane, adamantanone-2-oxime, 2-isonitrosoadamantane, 2-hydroxyimino adamantane, n-2-adamantylidene hydroxylamine, n-adamantan-2-ylidene hydroxylamine
N-(2-adamantylidene)hydroxylamine

Specifications

4500-12-3
White
MFCD00078273
2-adamantanone oxime, adamantan-2-one oxime, adamantanone oxime, 2-adamantanone, oxime, 2-hydroxyiminoadamantane, adamantanone-2-oxime, 2-isonitrosoadamantane, 2-hydroxyimino adamantane, n-2-adamantylidene hydroxylamine, n-adamantan-2-ylidene hydroxylamine
C1C2CC3CC1CC(C2)C3=NO
165.24
165.24
Crystalline Powder
168°C
C10H15NO
5 g
RABVIFXMFZFITE-KHPPLWFESA-N
N-(2-adamantylidene)hydroxylamine
64158
≥98.0% (GC)
2-Adamantanone Oxime
Videos
Safety and Handling

Safety and Handling

RTECSNumber : AU5018500

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'