missing translation for 'onlineSavingsMsg'
Learn More

2-Adamantanone Oxime 98.0+%, TCI America™

Catalog No. A21105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21105G 5 g
A211025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21105G Supplier TCI America Supplier No. A21105G
Only null left

Chemical Identifiers

CAS 4500-12-3
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.24
MDL Number MFCD00078273
InChI Key RABVIFXMFZFITE-KHPPLWFESA-N
Synonym 2-adamantanone oxime, adamantan-2-one oxime, adamantanone oxime, 2-adamantanone, oxime, 2-hydroxyiminoadamantane, adamantanone-2-oxime, 2-isonitrosoadamantane, 2-hydroxyimino adamantane, n-2-adamantylidene hydroxylamine, n-adamantan-2-ylidene hydroxylamine
PubChem CID 64158
IUPAC Name N-(2-adamantylidene)hydroxylamine
SMILES C1C2CC3CC1CC(C2)C3=NO

Specifications

Melting Point 168°C
Color White
Quantity 5 g
Formula Weight 165.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Adamantanone Oxime
RTECSNumber AU5018500
TSCA No