missing translation for 'onlineSavingsMsg'
Learn More

2-Adamantanone, 98%

CAS: 700-58-3 | C10H14O | 150.22 g/mol

$92.33 - $92.33

Chemical Identifiers

CAS 700-58-3
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00074737
InChI Key IYKFYARMMIESOX-UHFFFAOYSA-N
Synonym 2-adamantanone, adamantanone, 2-adamantone, 2-oxoadamantane, tricyclo 3.3.1.13,7 decanone, unii-ui7w503l08, tricyclo 3,3,1,13,7 decanone, tricyclo 3.3.1.1 sup 3,7 decanone, tricyclo 3.3.1.13,7 decan-2-one
PubChem CID 64151
ChEBI CHEBI:40611
IUPAC Name adamantan-2-one
SMILES C1C2CC3CC1CC(C2)C3=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC292500050
SDS View Documents
Thermo Scientific Chemicals
292500050
5 g Glass bottle
Each for $92.33
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

700-58-3
150.22
IYKFYARMMIESOX-UHFFFAOYSA-N
64151
adamantan-2-one
C10H14O
MFCD00074737
2-adamantanone, adamantanone, 2-adamantone, 2-oxoadamantane, tricyclo 3.3.1.13,7 decanone, unii-ui7w503l08, tricyclo 3,3,1,13,7 decanone, tricyclo 3.3.1.1 sup 3,7 decanone, tricyclo 3.3.1.13,7 decan-2-one
CHEBI:40611
C1C2CC3CC1CC(C2)C3=O

Specifications

700-58-3
100.0
White
97.5% min. (GC)
C10H14O
5 g
Solubility in water: insoluble. Other solubilities: soluble in methanol,ethanol and chloroform
C1C2CC3CC1CC(C2)C3=O
150.22
CHEBI:40611
98%
2-Adamantanone
97.5
282°C to 286°C
Authentic
Glass bottle
MFCD00074737
2-adamantanone, adamantanone, 2-adamantone, 2-oxoadamantane, tricyclo 3.3.1.13,7 decanone, unii-ui7w503l08, tricyclo 3,3,1,13,7 decanone, tricyclo 3.3.1.1 sup 3,7 decanone, tricyclo 3.3.1.13,7 decan-2-one
IYKFYARMMIESOX-UHFFFAOYSA-N
adamantan-2-one
64151
150.22
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-847-2

SDS
Documents

Documents

RUO – Research Use Only