missing translation for 'onlineSavingsMsg'
Learn More

2-Adamantanone, 98%

Catalog No. AC292501000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC292501000 100 g Glass bottle
AC292500050 5 g Glass bottle
AC292500250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC292501000 Supplier Thermo Scientific Chemicals Supplier No. 292501000

CAS: 700-58-3 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 700-58-3
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00074737
InChI Key IYKFYARMMIESOX-UHFFFAOYSA-N
Synonym 2-adamantanone, adamantanone, 2-adamantone, 2-oxoadamantane, tricyclo 3.3.1.13,7 decanone, unii-ui7w503l08, tricyclo 3,3,1,13,7 decanone, tricyclo 3.3.1.1 sup 3,7 decanone, tricyclo 3.3.1.13,7 decan-2-one
PubChem CID 64151
ChEBI CHEBI:40611
IUPAC Name adamantan-2-one
SMILES C1C2CC3CC1CC(C2)C3=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 282°C to 286°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol,ethanol and chloroform
Formula Weight 150.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Adamantanone
Show More Show Less
EINECSNumber 211-847-2

RUO – Research Use Only