missing translation for 'onlineSavingsMsg'
Learn More

2-Adamantanone, 98%

CAS: 700-58-3 | C10H14O | 150.221 g/mol

$92.85 - $863.23

Chemical Identifiers

CAS 700-58-3
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00074737
InChI Key IYKFYARMMIESOX-UHFFFAOYSA-N
Synonym 2-adamantanone, adamantanone, 2-adamantone, 2-oxoadamantane, tricyclo 3.3.1.13,7 decanone, unii-ui7w503l08, tricyclo 3,3,1,13,7 decanone, tricyclo 3.3.1.1 sup 3,7 decanone, tricyclo 3.3.1.13,7 decan-2-one
PubChem CID 64151
ChEBI CHEBI:40611
IUPAC Name adamantan-2-one
SMILES C1C2CC3CC1CC(C2)C3=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1427506
View Documents
Thermo Scientific Chemicals
A1427506
5 g
Each for $92.85
Only null left
 
AAA1427514
View Documents
Thermo Scientific Chemicals
A1427514
25 g
Each for $284.22
Only null left
 
AAA1427522
View Documents
Thermo Scientific Chemicals
A1427522
100 g
Each for $863.23
Only null left
 
Description

Description

It is used in the synthesis of dispiro N-Boc-protected 1,2,4-trioxane1 and (±)-1-(adamantan-2-yl)-2-propanamine. It is employed in reductive coupling (TiCl3/Li) which gives (adamantylidene)adamantane, an example of a general method for the synthesis of highly-substituted alkenes and in the preparation of highly-substituted alkenes by the Wittig reaction which gives poor yields with adamantanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of dispiro N-Boc-protected 1,2,4-trioxane1 and (+/-)-1-(adamantan-2-yl)-2-propanamine. It is employed in reductive coupling (TiCl3/Li) which gives (adamantylidene)adamantane, an example of a general method for the synthesis of highly-substituted alkenes and in the preparation of highly-substituted alkenes by the Wittig reaction which gives poor yields with adamantanone.

Solubility
Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

700-58-3
150.221
IYKFYARMMIESOX-UHFFFAOYSA-N
64151
adamantan-2-one
C10H14O
MFCD00074737
2-adamantanone, adamantanone, 2-adamantone, 2-oxoadamantane, tricyclo 3.3.1.13,7 decanone, unii-ui7w503l08, tricyclo 3,3,1,13,7 decanone, tricyclo 3.3.1.1 sup 3,7 decanone, tricyclo 3.3.1.13,7 decan-2-one
CHEBI:40611
C1C2CC3CC1CC(C2)C3=O

Specifications

700-58-3
C10H14O
5 g
2-adamantanone, adamantanone, 2-adamantone, 2-oxoadamantane, tricyclo 3.3.1.13,7 decanone, unii-ui7w503l08, tricyclo 3,3,1,13,7 decanone, tricyclo 3.3.1.1 sup 3,7 decanone, tricyclo 3.3.1.13,7 decan-2-one
C1C2CC3CC1CC(C2)C3=O
150.221
CHEBI:40611
98%
∼257°C (sublimation)
MFCD00074737
1210235
IYKFYARMMIESOX-UHFFFAOYSA-N
adamantan-2-one
64151
150.22
2-Adamantanone
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-847-2

RTECSNumber : AU5018000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only