missing translation for 'onlineSavingsMsg'
Learn More

2-Adamantanamine Hydrochloride 99.0+%, TCI America™

Catalog No. p-7117023
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A07185G 5 g
A071825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A07185G Supplier TCI America Supplier No. A07185G

Chemical Identifiers

CAS 10523-68-9
Molecular Formula C10H18ClN
Molecular Weight (g/mol) 187.71
MDL Number MFCD00074743
InChI Key WLDWDRZITJEWRJ-UHFFFAOYSA-N
Synonym 2-adamantanamine hydrochloride, adamantan-2-amine hydrochloride, 2-aminoadamantane hydrochloride, 2-adamantylamine hydrochloride, 2-adamantanamine hcl, 2-adamantanamine, hydrochloride, adamantan-2-ylamine hydrochloride, 1r,3r,5r,7r-adamantan-2-amine hydrochloride, tricyclo 3.3.1.13.7 dec-2-ylamine hydrochloride, tricyclo 3.3.1.13,7 decan-2-amine, hydrochloride
PubChem CID 25331
IUPAC Name hydrogen adamantan-2-amine chloride
SMILES [H+].[Cl-].NC1C2CC3CC(C2)CC1C3

Specifications

Melting Point 280°C
Color White-Yellow
Quantity 5 g
Formula Weight 187.71
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Adamantanamine Hydrochloride
RTECSNumber AU4375100
TSCA No