missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Adamantanamine hydrochloride, 99%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Glass bottle |
| Quantity | 5g |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in methanol and ethanol |
| Formula Weight | 187.71 |
| Percent Purity | 99% |
| Chemical Name or Material | 2-Adamantanamine hydrochloride |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement May cause respiratory irritation. Causes serious eye irritation. Causes skin irritation. |
| Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue |
| EINECSNumber | 234-074-2 |
RUO – Research Use Only