Learn More
2-Adamantanamine hydrochloride, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 167300250
| Quantity | 25g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 10523-68-9 | |
| 187.71 | |
| WLDWDRZITJEWRJ-UHFFFAOYSA-N | |
| 25331 | |
| [H+].[Cl-].NC1C2CC3CC(C2)CC1C3 |
| C10H18ClN | |
| MFCD00074743 | |
| 2-adamantanamine hydrochloride, adamantan-2-amine hydrochloride, 2-aminoadamantane hydrochloride, 2-adamantylamine hydrochloride, 2-adamantanamine hcl, 2-adamantanamine, hydrochloride, adamantan-2-ylamine hydrochloride, 1r,3r,5r,7r-adamantan-2-amine hydrochloride, tricyclo 3.3.1.13.7 dec-2-ylamine hydrochloride, tricyclo 3.3.1.13,7 decan-2-amine, hydrochloride | |
| adamantan-2-amine;hydrochloride |
Specifications
| 2-Adamantanamine hydrochloride | |
| 98.5 | |
| Authentic | |
| Glass bottle | |
| 25g | |
| 2-adamantanamine hydrochloride, adamantan-2-amine hydrochloride, 2-aminoadamantane hydrochloride, 2-adamantylamine hydrochloride, 2-adamantanamine hcl, 2-adamantanamine, hydrochloride, adamantan-2-ylamine hydrochloride, 1r,3r,5r,7r-adamantan-2-amine hydrochloride, tricyclo 3.3.1.13.7 dec-2-ylamine hydrochloride, tricyclo 3.3.1.13,7 decan-2-amine, hydrochloride | |
| WLDWDRZITJEWRJ-UHFFFAOYSA-N | |
| adamantan-2-amine;hydrochloride | |
| 25331 | |
| 99% |
| 10523-68-9 | |
| 100.0 | |
| 98.5% min. (GC) | |
| C10H18ClN | |
| MFCD00074743 | |
| Solubility in water: soluble. Other solubilities: soluble in methanol and ethanol | |
| [H+].[Cl-].NC1C2CC3CC(C2)CC1C3 | |
| 187.71 | |
| 187.71 |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue
GHS Signal Word: Warning
EINECSNumber : 234-074-2
Promotions