Learn More
2-Adamantanamine hydrochloride, 98+%
CAS: 10523-68-9 | C10H18ClN | 187.71 g/mol
$55.96 - $757.32
Chemical Identifiers
| CAS | 10523-68-9 |
|---|---|
| Molecular Formula | C10H18ClN |
| Molecular Weight (g/mol) | 187.71 |
| MDL Number | MFCD00074743 |
| InChI Key | WLDWDRZITJEWRJ-UHFFFAOYSA-N |
| Synonym | 2-adamantanamine hydrochloride, adamantan-2-amine hydrochloride, 2-aminoadamantane hydrochloride, 2-adamantylamine hydrochloride, 2-adamantanamine hcl, 2-adamantanamine, hydrochloride, adamantan-2-ylamine hydrochloride, 1r,3r,5r,7r-adamantan-2-amine hydrochloride, tricyclo 3.3.1.13.7 dec-2-ylamine hydrochloride, tricyclo 3.3.1.13,7 decan-2-amine, hydrochloride |
| PubChem CID | 25331 |
| SMILES | [H+].[Cl-].NC1C2CC3CC(C2)CC1C3 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1714003
|
Thermo Scientific Chemicals
A1714003 |
1 g |
Each for $55.96
|
|
|||||
|
AAA1714006
|
Thermo Scientific Chemicals
A1714006 |
5 g |
Each for $181.82
|
|
|||||
|
AAA1714014
|
Thermo Scientific Chemicals
A1714014 |
25 g |
Each for $757.32
|
|
|||||
Description
2-Adamantylamine hydrochloride was used to prepare 2-adamantylamide of 2?-(carboxymethoxime)-olivomycin I. The HPLC assay of 2-adamantylamine hydrochloride after pre-column derivatization with 4-fluoro-7-nitro-2,1,3-benzoxadiazole has been studied. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Adamantylamine hydrochloride was used to prepare 2-adamantylamide of 2′-(carboxymethoxime)-olivomycin I. The HPLC assay of 2-adamantylamine hydrochloride after pre-column derivatization with 4-fluoro-7-nitro-2,1,3-benzoxadiazole has been studied. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
Chemical Identifiers
| 10523-68-9 | |
| 187.71 | |
| WLDWDRZITJEWRJ-UHFFFAOYSA-N | |
| 25331 |
| C10H18ClN | |
| MFCD00074743 | |
| 2-adamantanamine hydrochloride, adamantan-2-amine hydrochloride, 2-aminoadamantane hydrochloride, 2-adamantylamine hydrochloride, 2-adamantanamine hcl, 2-adamantanamine, hydrochloride, adamantan-2-ylamine hydrochloride, 1r,3r,5r,7r-adamantan-2-amine hydrochloride, tricyclo 3.3.1.13.7 dec-2-ylamine hydrochloride, tricyclo 3.3.1.13,7 decan-2-amine, hydrochloride | |
| [H+].[Cl-].NC1C2CC3CC(C2)CC1C3 |
Specifications
| 10523-68-9 | |
| C10H18ClN | |
| 1 g | |
| 2-adamantanamine hydrochloride, adamantan-2-amine hydrochloride, 2-aminoadamantane hydrochloride, 2-adamantylamine hydrochloride, 2-adamantanamine hcl, 2-adamantanamine, hydrochloride, adamantan-2-ylamine hydrochloride, 1r,3r,5r,7r-adamantan-2-amine hydrochloride, tricyclo 3.3.1.13.7 dec-2-ylamine hydrochloride, tricyclo 3.3.1.13,7 decan-2-amine, hydrochloride | |
| WLDWDRZITJEWRJ-UHFFFAOYSA-N | |
| adamantan-2-amine;hydrochloride | |
| 25331 | |
| ≥98% |
| >300°C | |
| MFCD00074743 | |
| 4297901 | |
| Soluble in water. | |
| [H+].[Cl-].NC1C2CC3CC(C2)CC1C3 | |
| 187.71 | |
| 187.71 | |
| 2-Adamantanamine hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 234-074-2
RTECSNumber : AU4375100
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only