missing translation for 'onlineSavingsMsg'
Learn More

2-Adamantanamine hydrochloride, 98+%

Catalog No. AAA1714003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1714003 1 g
AAA1714006 5 g
AAA1714014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1714003 Supplier Thermo Scientific Chemicals Supplier No. A1714003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 10523-68-9 | C10H18ClN | 187.71 g/mol

2-Adamantylamine hydrochloride was used to prepare 2-adamantylamide of 2?-(carboxymethoxime)-olivomycin I. The HPLC assay of 2-adamantylamine hydrochloride after pre-column derivatization with 4-fluoro-7-nitro-2,1,3-benzoxadiazole has been studied. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Adamantylamine hydrochloride was used to prepare 2-adamantylamide of 2′-(carboxymethoxime)-olivomycin I. The HPLC assay of 2-adamantylamine hydrochloride after pre-column derivatization with 4-fluoro-7-nitro-2,1,3-benzoxadiazole has been studied. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.

Chemical Identifiers

CAS 10523-68-9
Molecular Formula C10H18ClN
Molecular Weight (g/mol) 187.71
MDL Number MFCD00074743
InChI Key WLDWDRZITJEWRJ-UHFFFAOYSA-N
Synonym 2-adamantanamine hydrochloride, adamantan-2-amine hydrochloride, 2-aminoadamantane hydrochloride, 2-adamantylamine hydrochloride, 2-adamantanamine hcl, 2-adamantanamine, hydrochloride, adamantan-2-ylamine hydrochloride, 1r,3r,5r,7r-adamantan-2-amine hydrochloride, tricyclo 3.3.1.13.7 dec-2-ylamine hydrochloride, tricyclo 3.3.1.13,7 decan-2-amine, hydrochloride
PubChem CID 25331
SMILES [H+].[Cl-].NC1C2CC3CC(C2)CC1C3

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 4297901
Solubility Information Soluble in water.
IUPAC Name adamantan-2-amine;hydrochloride
Formula Weight 187.71
Percent Purity ≥98%
Chemical Name or Material 2-Adamantanamine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 234-074-2
RTECSNumber AU4375100
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only