missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Acetylthiophene 98.0+%, TCI America™
$33.56 - $253.51
Chemical Identifiers
| CAS | 88-15-3 |
|---|---|
| Molecular Formula | C6H6OS |
| Molecular Weight (g/mol) | 126.173 |
| MDL Number | MFCD00005442 |
| InChI Key | WYJOVVXUZNRJQY-UHFFFAOYSA-N |
| Synonym | 2-acetylthiophene, 1-2-thienyl ethanone, 1-thiophen-2-yl-ethanone, methyl 2-thienyl ketone, 2-acetothienone, 2-acetothiophene, ethanone, 1-2-thienyl, 2-acetyl thiophene, ketone, methyl 2-thienyl, 2-thienyl methyl ketone |
| PubChem CID | 6920 |
| IUPAC Name | 1-thiophen-2-ylethanone |
| SMILES | CC(=O)C1=CC=CS1 |
Chemical Identifiers
| 88-15-3 | |
| 126.173 | |
| WYJOVVXUZNRJQY-UHFFFAOYSA-N | |
| 6920 | |
| CC(=O)C1=CC=CS1 |
| C6H6OS | |
| MFCD00005442 | |
| 2-acetylthiophene, 1-2-thienyl ethanone, 1-thiophen-2-yl-ethanone, methyl 2-thienyl ketone, 2-acetothienone, 2-acetothiophene, ethanone, 1-2-thienyl, 2-acetyl thiophene, ketone, methyl 2-thienyl, 2-thienyl methyl ketone | |
| 1-thiophen-2-ylethanone |
Specifications
| 88-15-3 | |
| 214°C | |
| MFCD00005442 | |
| 2-acetylthiophene, 1-2-thienyl ethanone, 1-thiophen-2-yl-ethanone, methyl 2-thienyl ketone, 2-acetothienone, 2-acetothiophene, ethanone, 1-2-thienyl, 2-acetyl thiophene, ketone, methyl 2-thienyl, 2-thienyl methyl ketone | |
| CC(=O)C1=CC=CS1 | |
| 126.173 | |
| 126.17 | |
| Liquid |
| Yellow | |
| C6H6OS | |
| 25 g | |
| WYJOVVXUZNRJQY-UHFFFAOYSA-N | |
| 1-thiophen-2-ylethanone | |
| 6920 | |
| ≥98.0% (GC) | |
| 2-Acetylthiophene |
Safety and Handling
RTECSNumber : OB6300000
TSCA : Yes