missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylthiophene 98.0+%, TCI America™

Catalog No. A072425G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A072425G 25 g
A0724100G 100 g
A0724500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A072425G Supplier TCI America Supplier No. A072425G

Chemical Identifiers

CAS 88-15-3
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.173
MDL Number MFCD00005442
InChI Key WYJOVVXUZNRJQY-UHFFFAOYSA-N
Synonym 2-acetylthiophene, 1-2-thienyl ethanone, 1-thiophen-2-yl-ethanone, methyl 2-thienyl ketone, 2-acetothienone, 2-acetothiophene, ethanone, 1-2-thienyl, 2-acetyl thiophene, ketone, methyl 2-thienyl, 2-thienyl methyl ketone
PubChem CID 6920
IUPAC Name 1-thiophen-2-ylethanone
SMILES CC(=O)C1=CC=CS1

Specifications

Color Yellow
Boiling Point 214°C
Quantity 25 g
Formula Weight 126.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Acetylthiophene
RTECSNumber OB6300000
TSCA Yes