missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylthiophene, 99%

CAS: 88-15-3 | C6H6OS | 126.173 g/mol

$100.45 - $333.64

Chemical Identifiers

CAS 88-15-3
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.173
MDL Number MFCD00005442
InChI Key WYJOVVXUZNRJQY-UHFFFAOYSA-N
Synonym 2-acetylthiophene, 1-2-thienyl ethanone, 1-thiophen-2-yl-ethanone, methyl 2-thienyl ketone, 2-acetothienone, 2-acetothiophene, ethanone, 1-2-thienyl, 2-acetyl thiophene, ketone, methyl 2-thienyl, 2-thienyl methyl ketone
PubChem CID 6920
IUPAC Name 1-thiophen-2-ylethanone
SMILES CC(=O)C1=CC=CS1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1306222
View Documents
Thermo Scientific Chemicals
A1306222
100 g
Each for $100.45
Only null left
 
AAA1306236
View Documents
Thermo Scientific Chemicals
A1306236
500 g
Each for $333.64
Only null left
 
Description

Description

2-Acetylthiophene is used as an intermediate in the manufacturing of drugs like Tiamonium Iodide, Suprofan, Stepronin, Tenonitrozole, Namirotene, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylthiophene is used as an intermediate in the manufacturing of drugs like Tiamonium Iodide, Suprofan, Stepronin, Tenonitrozole, Namirotene, etc.

Solubility
Insoluble in water.

Notes
It is sensitive to light. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

88-15-3
126.173
WYJOVVXUZNRJQY-UHFFFAOYSA-N
6920
CC(=O)C1=CC=CS1
C6H6OS
MFCD00005442
2-acetylthiophene, 1-2-thienyl ethanone, 1-thiophen-2-yl-ethanone, methyl 2-thienyl ketone, 2-acetothienone, 2-acetothiophene, ethanone, 1-2-thienyl, 2-acetyl thiophene, ketone, methyl 2-thienyl, 2-thienyl methyl ketone
1-thiophen-2-ylethanone

Specifications

88-15-3
1.168
91°C (195°F)
C6H6OS
MFCD00005442
UN2810
Light sensitive
Insoluble in water.
CC(=O)C1=CC=CS1
126.173
126.18
2-Acetylthiophene
10°C to 12°C
213°C to 214°C
Slightly Sweetish
1.566
100 g
107910
2-acetylthiophene, 1-2-thienyl ethanone, 1-thiophen-2-yl-ethanone, methyl 2-thienyl ketone, 2-acetothienone, 2-acetothiophene, ethanone, 1-2-thienyl, 2-acetyl thiophene, ketone, methyl 2-thienyl, 2-thienyl methyl ketone
WYJOVVXUZNRJQY-UHFFFAOYSA-N
1-thiophen-2-ylethanone
6920
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H311-H330-H227
Fatal if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Combustible liquid.

P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P370+P378q-P501c

H227-H301+H311+H331

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

EINECSNumber : 201-804-6

RTECSNumber : OB6300000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only