missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylthiophene, 99%

Catalog No. AAA1306236
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1306236 500 g
AAA1306222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1306236 Supplier Thermo Scientific Chemicals Supplier No. A1306236
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 88-15-3 | C6H6OS | 126.173 g/mol

2-Acetylthiophene is used as an intermediate in the manufacturing of drugs like Tiamonium Iodide, Suprofan, Stepronin, Tenonitrozole, Namirotene, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylthiophene is used as an intermediate in the manufacturing of drugs like Tiamonium Iodide, Suprofan, Stepronin, Tenonitrozole, Namirotene, etc.

Solubility
Insoluble in water.

Notes
It is sensitive to light. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 88-15-3
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.173
MDL Number MFCD00005442
InChI Key WYJOVVXUZNRJQY-UHFFFAOYSA-N
Synonym 2-acetylthiophene, 1-2-thienyl ethanone, 1-thiophen-2-yl-ethanone, methyl 2-thienyl ketone, 2-acetothienone, 2-acetothiophene, ethanone, 1-2-thienyl, 2-acetyl thiophene, ketone, methyl 2-thienyl, 2-thienyl methyl ketone
PubChem CID 6920
IUPAC Name 1-thiophen-2-ylethanone
SMILES CC(=O)C1=CC=CS1

Specifications

Melting Point 10°C to 12°C
Density 1.168
Boiling Point 213°C to 214°C
Flash Point 91°C (195°F)
Odor Slightly Sweetish
Refractive Index 1.566
Quantity 500 g
UN Number UN2810
Beilstein 107910
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 126.18
Percent Purity 99%
Chemical Name or Material 2-Acetylthiophene
Show More Show Less
Hazard Category H227-H301+H311+H331
Hazard Statement GHS H Statement
H300-H311-H330-H227
Fatal if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 201-804-6
RTECSNumber OB6300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only