missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylthiophene, 98%

CAS: 88-15-3 | C6H6OS | 126.17 g/mol

$97.48 - $364.09

Chemical Identifiers

CAS 88-15-3
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.17
MDL Number MFCD00005442
InChI Key WYJOVVXUZNRJQY-UHFFFAOYSA-N
Synonym 2-acetylthiophene, 1-2-thienyl ethanone, 1-thiophen-2-yl-ethanone, methyl 2-thienyl ketone, 2-acetothienone, 2-acetothiophene, ethanone, 1-2-thienyl, 2-acetyl thiophene, ketone, methyl 2-thienyl, 2-thienyl methyl ketone
PubChem CID 6920
IUPAC Name 1-thiophen-2-ylethanone
SMILES CC(=O)C1=CC=CS1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC102721000
SDS View Documents
Thermo Scientific Chemicals
102721000
100 g Glass bottle
Each for $97.48
Only null left
 
AC102725000
SDS View Documents
Thermo Scientific Chemicals
102725000
500 g Glass bottle
Each for $364.09
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

88-15-3
126.17
WYJOVVXUZNRJQY-UHFFFAOYSA-N
6920
CC(=O)C1=CC=CS1
C6H6OS
MFCD00005442
2-acetylthiophene, 1-2-thienyl ethanone, 1-thiophen-2-yl-ethanone, methyl 2-thienyl ketone, 2-acetothienone, 2-acetothiophene, ethanone, 1-2-thienyl, 2-acetyl thiophene, ketone, methyl 2-thienyl, 2-thienyl methyl ketone
1-thiophen-2-ylethanone

Specifications

88-15-3
100.0
Yellow to Brown
214°C
Authentic
Glass bottle
1.5640 to 1.566
MFCD00005442
17, 287
2-acetylthiophene, 1-2-thienyl ethanone, 1-thiophen-2-yl-ethanone, methyl 2-thienyl ketone, 2-acetothienone, 2-acetothiophene, ethanone, 1-2-thienyl, 2-acetyl thiophene, ketone, methyl 2-thienyl, 2-thienyl methyl ketone
WYJOVVXUZNRJQY-UHFFFAOYSA-N
1-thiophen-2-ylethanone
6920
98%
2-Acetylthiophene, 98%
97.5
10°C to 11°C
1.1600g/mL
91°C
97.5% min. (GC)
C6H6OS
(C4H3S)COCH3
100 g
1.16
Solubility in water: 14g/L (30°C). Other solubilities: soluble in ethanol,benzene,chloroform,benzene
CC(=O)C1=CC=CS1
126.17
126.17
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-804-6

RTECSNumber : OB6300000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only