missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylthiazole 98.0+%, TCI America™

Catalog No. p-7117337
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12655G 5 g
A126525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A12655G Supplier TCI America Supplier No. A12655G
Only null left

Chemical Identifiers

CAS 24295-03-2
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.161
MDL Number MFCD00005324
InChI Key MOMFXATYAINJML-UHFFFAOYSA-N
Synonym 2-acetylthiazole, 1-1,3-thiazol-2-yl ethan-1-one, ethanone, 1-2-thiazolyl, 1-1,3-thiazol-2-yl ethanone, 2-acetyl thiazole, 1-2-thiazolyl ethanone, ketone, methyl 2-thiazolyl, 2-acetylthiazol, 2-acetyl-1,3-thiazole, 1-thiazol-2-yl-ethanone
PubChem CID 520108
IUPAC Name 1-(1,3-thiazol-2-yl)ethanone
SMILES CC(=O)C1=NC=CS1

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
Formula Weight 127.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Acetylthiazole