missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylthiazole, 99%

Catalog No. AAA1126603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1126603 1 g
AAA1126606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1126603 Supplier Thermo Scientific Chemicals Supplier No. A1126603

CAS: 24295-03-2 | C5H5NOS | 127.161 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24295-03-2
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.161
MDL Number MFCD00005324
InChI Key MOMFXATYAINJML-UHFFFAOYSA-N
Synonym 2-acetylthiazole, 1-1,3-thiazol-2-yl ethan-1-one, ethanone, 1-2-thiazolyl, 1-1,3-thiazol-2-yl ethanone, 2-acetyl thiazole, 1-2-thiazolyl ethanone, ketone, methyl 2-thiazolyl, 2-acetylthiazol, 2-acetyl-1,3-thiazole, 1-thiazol-2-yl-ethanone
PubChem CID 520108
IUPAC Name 1-(1,3-thiazol-2-yl)ethanone
SMILES CC(=O)C1=NC=CS1

Specifications

Density 1.227
Boiling Point 89°C to 91°C (12 mmHg)
Flash Point 78°C (172°F)
Odor Characteristic
Refractive Index 1.548
Quantity 1 g
Beilstein 109803
Formula Weight 127.17
Percent Purity 99%
Chemical Name or Material 2-Acetylthiazole
Hazard Category H227-H302
Hazard Statement GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 246-134-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only